1,1'-Bis(diisopropylphosphino)ferrocene, Sigma-Aldrich®
Catalog # SIAL578266-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:410.25 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:97239-80-0
Formula:C₂₂H₂₈FeP₂
MDL Number:MFCD02684559