Order Entry
Poland
ContactUsLinkComponent
1,1'-Bis(di-tert-butylphosphino)ferrocene, Sigma-Aldrich®
1,1'-Bis(di-tert-butylphosphino)ferrocene, Sigma-Aldrich®
Catalog # SIAL695149-1G
Supplier:  Merck
CAS Number:  
1,1'-Bis(di-tert-butylphosphino)ferrocene, Sigma-Aldrich®
Catalog # SIAL695149-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :474.42 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:84680-95-5
Formula:C₂₆H₄₄FeP₂
MDL Number:MFCD01630818