Order Entry
Poland
ContactUsLinkComponent
Safety spectacles, KLEENGUARD* V20
Safety spectacles, KLEENGUARD* V20
Catalog # KIMB25656
Supplier:  KIMBERLY CLARK
undefined
Safety spectacles, KLEENGUARD* V20
Catalog # KIMB25656
Supplier:  KIMBERLY CLARK
Specifications
  • Type:
    Anti-UV
  • Colour:
    Clear
  • Seq:
    04
  • Lenses:
    Indoor/Outdoor
  • Pk:
    1
Specifications
About this item
A stylish new standard in eye protection that improves personal comfort, safety and worker performance. KLEENGUARD* V20 Comfort Eye Protection features distortion-free lenses with proven impact protection, added comfort and a broad range of styles.
  • 99,9% UVA/UVB protection
  • Wrap-around lens for better eye coverage
  • Comfortable, padded temples
Certifications: EN 166; 1F (111-9010: 1FT); ANSI Z87.1, CSA Z94.3