Order Entry
Poland
ContactUsLinkComponent
Crystal violet solution Reag. Ph. Eur. 1022901
Crystal violet solution Reag. Ph. Eur. 1022901
Catalog # 87811.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Crystal violet solution Reag. Ph. Eur. 1022901
Catalog # 87811.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :407.99 g/mol
UN:2789
ADR:8 (3),II
 :GHS02
 :GHS05
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750