4-Mercaptobenzoic acid, Sigma-Aldrich®
Catalog # SIAL706329-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.1863 g/mol
CAS Number:1074-36-8
Formula:C₇H₆O₂S
MDL Number:MFCD00016617