Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Chlorophenyl)ethyl isothiocyanate
2-(4-Chlorophenyl)ethyl isothiocyanate, Sigma-Aldrich®
2-(4-Chlorophenyl)ethyl isothiocyanate, Sigma-Aldrich®
2-(4-Chlorophenyl)ethyl isothiocyanate, Sigma-Aldrich®
SIAL591378-5G
:
Merck
:
2-(4-Chlorophenyl)ethyl isothiocyanate, Sigma-Aldrich®
SIAL591378-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
M-Clarity™ quality level = MQ100
:
197.69 g/mol
:
17608-10-5
:
C₉H₈ClNS
:
MFCD00041127