Order Entry
Poland
Orders LinkContactUsLinkComponent
Capillary columns, TRACE™ TR-1
Capillary columns, TRACE™ TR-1
Capillary columns, TRACE™ TR-1
Certificates
Capillary columns, TRACE™ TR-1
Capillary columns, TRACE™ TR-1
14 Options Available Below
Product Details & Documents
TRACE™ TR-1 is a general purpose 100% dimethylpolysiloxane non-polar column suitable for a wide range of compounds, including hydrocarbons, esters, pesticides, drugs and amines.
  • Low bleed for increased sensitivity
  • High operating temperature for easy reconditioning
  • Solvent washable
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TR-1
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 240 zł
    4 240 zł
    1 * 1 SZT
       4 240 zł
  • Capillary
    TR-1
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 330 zł
    2 330 zł
    1 * 1 SZT
       2 330 zł
  • Capillary
    TR-1
    0,25 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 470 zł
    2 470 zł
    1 * 1 SZT
       2 470 zł
  • Capillary
    TR-1
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 3 830 zł
    3 830 zł
    1 * 1 SZT
       3 830 zł
  • Capillary
    TR-1
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 080 zł
    4 080 zł
    1 * 1 SZT
       4 080 zł
  • Capillary
    TR-1
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 730 zł
    6 730 zł
    1 * 1 SZT
       6 730 zł
  • Capillary
    TR-1
    0,50 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 3 940 zł
    3 940 zł
    1 * 1 SZT
       3 940 zł
  • Capillary
    TR-1
    0,50 µm
    0,25 mm
    100 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 8 060 zł
    8 060 zł
    1 * 1 SZT
       8 060 zł
  • Capillary
    TR-1
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 3 990 zł
    3 990 zł
    1 * 1 SZT
       3 990 zł
  • Capillary
    TR-1
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 240 zł
    4 240 zł
    1 * 1 SZT
       4 240 zł
  • Capillary
    TR-1
    1,00 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 730 zł
    6 730 zł
    1 * 1 SZT
       6 730 zł
  • Capillary
    TR-1
    3,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 240 zł
    4 240 zł
    1 * 1 SZT
       4 240 zł
  • Capillary
    TR-1
    5,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 190 zł
    4 190 zł
    1 * 1 SZT
       4 190 zł
  • Capillary
    TR-1
    5,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 410 zł
    4 410 zł
    1 * 1 SZT
       4 410 zł