Order Entry
Poland
ContactUsLinkComponent
 
1-Pyrenebutyric hydrazide, Sigma-Aldrich®
Catalog # SIAL82669-100MG
Supplier:  Merck
CAS Number:  
1-Pyrenebutyric hydrazide, Sigma-Aldrich®
Catalog # SIAL82669-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :302.3697 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:55486-13-0
Formula:C₂₀H₁₈N₂O
MDL Number:MFCD00467260