HPLC columns, Hypersil GOLD™ SAX
Supplier: Thermo Fisher Scientific
Certificates
About this item
These quaternary amine strong anion-exchange columns are highly stable for aqueous and low pH mobile phases.
- Analyse smaller organic molecules, including nucleotides and organic acids
- Outstanding peak shape and sensitivity
Based on high purity silica bonded with an ion-exchange ligand, Hypersil GOLD™ SAX HPLC columns provide outstanding peak shape and sensitivity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2,1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4,6 mm int.Ø require the 4/4,6 mm UNIGUARD holder (10046-744).
Specifications
- Catalog No:
- 555-6530
- 555-6518
- 555-6396
- 554-4329
- 554-4331
- 555-6380
- 554-4330
- 555-6516
- 555-6401
- 555-6381
- Carbon load:
- 2,50%
- 2,50%
- 2,50%
- 2,50%
- 2,50%
- 2,50%
- 2,50%
- 2,50%
- 2,50%
- pH range:
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- 2 to 8
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g