cis-3-Hexenyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3681-71-8
:
Formula:C₈H₁₄O₂
MDL Number:MFCD00036563
:142.20 g/mol
ADR:3,III
:M-Clarity™ quality level = MQ300
EINECS:222-960-1
UN:3272
Frequently Bought Together