Order Entry
Poland
ContactUsLinkComponent
Benzyldimethyltetradecylammonium chloride hydrate ≥98%
Corrosive
Irritant
Benzyldimethyltetradecylammonium chloride hydrate ≥98%
Catalog # J63465.14
CAS Number:  
Corrosive
Irritant
Benzyldimethyltetradecylammonium chloride hydrate ≥98%
Catalog # J63465.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :368.05 g/mol (anhy)
Merck Index:14,1059
Storage Temperature:Ambient
EINECS:205-352-0
UN:2923
ADR:8,III
 :GHS05
 :GHS07
CAS Number:139-08-2
Formula:C₂₃H₄₂ClN·H₂O
MDL Number:MFCD00011771