1,1'-Bis(diphenylphosphino)ferrocene, Sigma-Aldrich®
Catalog # SIAL555088-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:554.39 g/mol
:M-Clarity™ quality level = MQ100
EINECS:430-420-3
CAS Number:12150-46-8
Formula:C₃₄H₂₈FeP₂
MDL Number:MFCD00001422