Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hydrogen carbonate
Sodium hydrogen carbonate 5 g/l in aqueous solution, pure
Sodium hydrogen carbonate 5 g/l in aqueous solution, pure
CHMP428105307.1000
:
CHEMPUR
:
Sodium hydrogen carbonate 5 g/l in aqueous solution, pure
CHMP428105307.1000
:
CHEMPUR
:
Pack type:
Glass bottle
Pk:
1 L
:
:
84.01 g/mol
:
Ambient
:
205-633-8
:
144-55-8
:
NaHCO₃
:
MFCD00003528