Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium dichromate
Potassium dichromate 1/60 mol concentrated aqueous solution
Potassium dichromate 1/60 mol concentrated aqueous solution
Potassium dichromate 1/60 mol concentrated aqueous solution
CHMP167411009.0001
:
CHEMPUR
:
Potassium dichromate 1/60 mol concentrated aqueous solution
CHMP167411009.0001
:
CHEMPUR
:
Pk:
1
:
:
294.18 g/mol
:
Ambient
:
231-906-6
:
3287
:
6.1,III
:
GHS06
:
GHS08
:
7778-50-9
:
K₂Cr₂O₇
:
MFCD00011367