Order Entry
Poland
ContactUsLinkComponent
Trimethyltin chloride 1 mol/l in hexane, Sigma-Aldrich® | Trimethyltin chloride
Trimethyltin chloride 1 mol/l in hexane, Sigma-Aldrich®
Catalog # SIAL375187-100ML
Supplier:  Merck
CAS Number:  
Trimethyltin chloride 1 mol/l in hexane, Sigma-Aldrich®
Catalog # SIAL375187-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :199.27 g/mol
ADR:3,II
 :M-Clarity™ quality level = MQ100
EINECS:213-917-8
UN:1992
CAS Number:1066-45-1
Formula:C₃H₉ClSn
MDL Number:MFCD00000520