Tetrabutylphosphonium hydroxide 40% roztwór wodny, Sigma-Aldrich®
Catalog # SIAL438294-100ML
Supplier: Merck
CAS Number:
Specifications
- Rodzaj opakowania:Glass bottle
- Ilość:100 mL
- Seq:2440
Specifications
About this item
:
:276.45 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:238-528-0
UN:3267
CAS Number:14518-69-5
Formula:C₁₆H₃₇OP
MDL Number:MFCD00068456