
Tetrabutylphosphonium hydroxide 40% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14518-69-5
:
Formula:C₁₆H₃₇OP
MDL Number:MFCD00068456
:276.45 g/mol
Density:0.989
Storage Temperature:Ambient
EINECS:238-528-0
UN:3267
ADR:8,II
:GHS05
Related Information
Frequently Bought Together