Tris(pentafluorophenyl)borane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1109-15-5
:
Formula:C₁₈BF₁₅
MDL Number:MFCD00269813
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:511.98 g/mol
EINECS:213-504-2
UN:2811
Frequently Bought Together