
2-Amino-4'-bromoacetophenone hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5467-72-1
:
Formula:C₈H₉BrClNO
MDL Number:MFCD00051998
:250.53 g/mol
Melting Pt:280…283 °C
EINECS:226-778-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together