Order Entry
Poland
ContactUsLinkComponent
1,2-Dibromoethane-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
1,2-Dibromoethane-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487198-1G
Supplier:  Merck
CAS Number:  
1,2-Dibromoethane-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL487198-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:33458-49-0
Formula:C₂H₄Br₂
MDL Number:MFCD00142466