Order Entry
Poland
ContactUsLinkComponent
6-O-Palmitoyl-L-ascorbic acid
6-O-Palmitoyl-L-ascorbic acid
Catalog # MOLE43238027-100G
Supplier:  Molekula
CAS Number:  
6-O-Palmitoyl-L-ascorbic acid
Catalog # MOLE43238027-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :414.54 g/mol
Boiling Pt:∼250 °C
Melting Pt:107…117 °C
EINECS:205-305-4
CAS Number:137-66-6
Formula:C₂₂H₃₈O₇
MDL Number:MFCD00005377