Order Entry
Poland
Orders LinkContactUsLinkComponent
6-O-Palmitoyl-L-ascorbic acid ≥99%

6-O-Palmitoyl-L-ascorbic acid ≥99%

6-O-Palmitoyl-L-ascorbic acid ≥99%
Certificates
6-O-Palmitoyl-L-ascorbic acid ≥99%
6-O-Palmitoyl-L-ascorbic acid ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 137-66-6 Molecular Formula: C22H38O7 Formula Weight: 414.52 Physical Form: Crystalline Odour: Pleasant MDL No.: MFCD00005377

Synonyms:  L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate
  • CAS Number:137-66-6
  • Formula:C₂₂H₃₈O₇
  • MDL Number:MFCD00005377
  • Molecular Weight:414.54 g/mol
  • Boiling Pt:∼250 °C
  • Melting Pt:113…114 °C
  • Storage Temperature:Ambient
  • EINECS:205-305-4
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.