Order Entry
Poland
ContactUsLinkComponent
5,7-Hexadecadiynoic acid, Sigma-Aldrich®
5,7-Hexadecadiynoic acid, Sigma-Aldrich®
Catalog # SIAL544027-1G
Supplier:  Merck
CAS Number:  
5,7-Hexadecadiynoic acid, Sigma-Aldrich®
Catalog # SIAL544027-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
CAS Number:28393-01-3
Formula:C₁₆H₂₄O₂
MDL Number:MFCD03695477