Order Entry
Poland
Orders LinkContactUsLinkComponent
 

(S)-(+)-2,2-Dimethylcyclopropanecarboxamide

Glass bottle for solids, 5 g
Catalog # MOLE17190958-5G
Marketsource
Supplier:   Molekula
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide
Catalog # MOLE17190958-5G
Supplier:  Molekula
Marketsource
CAS Number:  75885-58-4
(S)-(+)-2,2-Dimethylcyclopropanecarboxamide

Glass bottle for solids, 5 g

CAS Number:  75885-58-4
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (S)-2,2-Dimethylcyclopropanecarboxamide
  • CAS Number:75885-58-4
  • Formula:C₆H₁₁NO
  • MDL Number:MFCD00216614
  • Molecular Weight:113.1600 g/mol
  • EINECS:278-334-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications