Order Entry
Poland
ContactUsLinkComponent
Buffers, acetate, 1M solutions, pH 3,0 to 5,5
Buffers, acetate, 1M solutions, pH 3,0 to 5,5
Certificates
Buffers, acetate, 1M solutions, pH 3,0 to 5,5
Certificates

Specifications

  • CAS No.:
    127-09-3
  • MDL No.:
    MFCD00012459
  • Physical form:
    Liquid

Specifications

Frequently Bought Together