Order Entry
Poland
ContactUsLinkComponent
Sodium-D(+)-glucuronate monohydrate for synthesis, Sigma-Aldrich®
Sodium-D(+)-glucuronate monohydrate for synthesis, Sigma-Aldrich®
Catalog # 8.14723.0010
Supplier:  Merck
CAS Number:  
Sodium-D(+)-glucuronate monohydrate for synthesis, Sigma-Aldrich®
Catalog # 8.14723.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    137 - 141 °C
  • Molar Mass:
    234.14 g/mol

Specification Test Results

About this item

 :
 :234.14 g/mol (216.14 anhy)
 :M-Clarity™ quality level = MQ200
EINECS:239-065-7
CAS Number:207300-70-7
Formula:C₆H₁₀O₇·1H₂O
MDL Number:MFCD00151051