
N-Boc-glycine (boc-gly-OH) 99+%
:4530-20-5
:
:C₇H₁₃NO₄
:MFCD00002690
:175.2 g/mol
:87…90 °C
:224-864-5
:GHS05
:Refrigerator
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:85 to 90 °C
- Water:≤0.5 % (K.F.)
- HPLC:≥99.0 % (ELSD)
- Titration with NaOH:≥99.0 %
- Heavy metals (as Pb):≤5 ppm
Frequently Bought Together