

Indium(III) isopropoxide 5% (w/v) in 2-propanol , 99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38218-24-5
:
Formula:C₉H₂₁InO₃
MDL Number:MFCD00210318
:292.08 g/mol
Boiling Pt:82 °C
Density:0.78
Flash Pt:12 °C (54 °F)
Storage Temperature:Ambient
UN:1219
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together