Order Entry
Poland
ContactUsLinkComponent
Magnesium chloride, pure
undefined
Magnesium chloride, pure
  • Appearance:
    White Powder (May contain grey to black particles)
  • Titration Complexometric:
    ≥98.0 % (on dry substance)
  • Water:
    ≤3 % (K.F.)
Frequently Bought Together