


Methanol for HPLC, general purpose reagent
Catalog # CHMP616219905.2500
Supplier: CHEMPUR
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
About this item
:
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595