Potassium iodide 1 mol/l (1 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Storage Temperature:Ambient
EINECS:231-659-4
Frequently Bought Together