Order Entry
Poland
ContactUsLinkComponent
3-Bromobenzylamine hydrochloride
3-Bromobenzylamine hydrochloride
Catalog # MOLEM68181301
Supplier:  Molekula
CAS Number:  
3-Bromobenzylamine hydrochloride
Catalog # MOLEM68181301
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :222.52 g/mol
CAS Number:39959-54-1
Formula:C₇H₉BrClN
MDL Number:MFCD00012856