Order Entry
Poland
ContactUsLinkComponent
2-Bromobenzylamine hydrochloride, Sigma-Aldrich®
2-Bromobenzylamine hydrochloride, Sigma-Aldrich®
Catalog # SIAL225169-5G
Supplier:  Merck
CAS Number:  
2-Bromobenzylamine hydrochloride, Sigma-Aldrich®
Catalog # SIAL225169-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :222.52 g/mol
CAS Number:5465-63-4
Formula:C₇H₉BrClN
MDL Number:MFCD00012853