Potassium iodide 0.1 mol concentrated aqueous solution volumetric solution
Catalog # CHMP167431800.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:166 g/mol
Storage Temperature:Ambient
EINECS:231-659-4
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405