


N,N-Dimethylformamide-[¹⁵N] (99% ¹⁵N), Sigma-Aldrich®
Catalog # SIAL493074-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
ADR:3,III
:M-Clarity™ quality level = MQ200
UN:2265
:GHS02
:GHS08
:GHS07
CAS Number:1449-84-9
Formula:HCOON(CH₃)₂