Złóż zamówienie
Poland
ContactUsLinkComponent
Methyl tetrahydropyran-4-carboxylate
Methyl tetrahydropyran-4-carboxylate
Catalog # MOLEM78224733
Supplier:  Molekula
CAS Number:  
Methyl tetrahydropyran-4-carboxylate
Catalog # MOLEM78224733
Supplier:  Molekula
CAS Number:  

Specifications

  • Rodzaj opakowania:
    Glass bottle for solids
  • Ilość:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :144.1684 g/mol
CAS Number:110238-91-0
Formula:C₇H₁₂O₃
MDL Number:MFCD01075170