Order Entry
Poland
ContactUsLinkComponent
Spectrocolorimeter, LC 100, Lovibond®
Spectrocolorimeter, LC 100, Lovibond®
Certificates
undefined
Spectrocolorimeter, LC 100, Lovibond®
Specifications
  • Catalog No:
  • 633-0243
  • 633-0242
  • Model:
  • LC 100
  • LC 100
  • Illuminants:
  • D65
  • D65
  • Light source:
  • 25 independent tridirectional LEDs
  • 25 independent tridirectional LEDs
  • Measuring area:
  • 4 + 8 mm
  • 4 + 8 mm
  • Observer:
  • A (10)
  • A (10)
  • Measuring geometry:
  • 45/0 image capture
  • 45/0 image capture
  • Reproducibility:
  • short term typically 0,10 ΔE 94 on white (d65/10)
  • short term typically 0,10 ΔE 94 on white (d65/10)
  • Measurement time:
  • 1,8 s
  • 1,8 s
  • Display:
  • 45 mm colour TFT
  • 45 mm colour TFT
  • Storable data sets:
  • 20 standards/350 samples
  • 20 standards/350 samples
  • Interfaces:
  • USB
  • USB