Order Entry
Poland
ContactUsLinkComponent
Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid with hydrofluoric acid (max. 1%) standard for ICP
Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid with hydrofluoric acid (max. 1%) standard for ICP
Supplier:  CPACHEM
Certificates
Zirconium standard solution, 1000 mg/L Zr in 5% hydrochloric acid with hydrofluoric acid (max. 1%) standard for ICP
Supplier:  CPACHEM