Potassium dichromate, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7778-50-9
:
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367
:294.18 g/mol
Merck Index:12,07790
Boiling Pt:500 °C (1013 hPa)
Melting Pt:398 °C
Density:2.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-906-6
UN:3087
ADR:5.1,II
REACH:01-2119454792-32
:GHS03
:GHS06
:GHS05
:GHS08
:GHS09
Frequently Bought Together