Magnesium chloride 0.1 mol/l (0.2 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7786-30-3
:
Formula:MgCl₂
MDL Number:MFCD00011106
:95.21 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
Frequently Bought Together