Order Entry
Poland
ContactUsLinkComponent
Zinc sulphate 0.01 mol/l (0.01 N) in aqueous solution volumetric solution
Zinc sulphate 0.01 mol/l (0.01 N) in aqueous solution volumetric solution
Catalog # POCH265751161-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  
Zinc sulphate 0.01 mol/l (0.01 N) in aqueous solution volumetric solution
Catalog # POCH265751161-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302