Zinc sulphate 0.01 mol/l (0.01 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7733-02-0
:
Formula:O₄SZn
MDL Number:MFCD00011302
:161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
Frequently Bought Together