Magnesium, turnings
Supplier: CHEMPUR
Jump to:
About this item
- CAS Number:7439-95-4
- Formula:Mg
- MDL Number:MFCD00085308
- Molecular Weight:24.3 g/mol
- Storage Temperature:Ambient
- EINECS:231-104-6
5 Options Available Below
Product Details & Documents
- CAS Number:7439-95-4
- Formula:Mg
- MDL Number:MFCD00085308
- Molecular Weight:24.3 g/mol
- Storage Temperature:Ambient
- EINECS:231-104-6
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



