Order Entry
Poland
ContactUsLinkComponent
 
Di-n-octyl phthalate-[ring-D₄] (98% D), Sigma-Aldrich®
Catalog # SIAL488887-100MG
Supplier:  Merck
CAS Number:  
Di-n-octyl phthalate-[ring-D₄] (98% D), Sigma-Aldrich®
Catalog # SIAL488887-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
ADR:9,III
 :M-Clarity™ quality level = MQ200
UN:3082
CAS Number:93952-13-7
Formula:C₂₄H₃₄D₄O₄
MDL Number:MFCD00144150