Bis(2-ethylhexyl) phthalate-[ring-D₄] (98% D), Sigma-Aldrich®
Catalog # SIAL617180-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:93951-87-2
Formula:C₂₄H₃₄D₄O₄
MDL Number:MFCD00143759