Bis(2-ethylhexyl) phthalate-[ring-D₄] (98% D)
Catalog # PROOCILDLM13680.25
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:93951-87-2
Formula:C₂₄H₃₄D₄O₄
MDL Number:MFCD00143759