Order Entry
Poland
ContactUsLinkComponent
N-Ethyldiisopropylamine 2 mol/l in N-methyl-2-pyrrolidone, Sigma-Aldrich®
N-Ethyldiisopropylamine 2 mol/l in N-methyl-2-pyrrolidone, Sigma-Aldrich®
Catalog # SIAL03439-250ML
Supplier:  Merck
CAS Number:  
N-Ethyldiisopropylamine 2 mol/l in N-methyl-2-pyrrolidone, Sigma-Aldrich®
Catalog # SIAL03439-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :129.25 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:230-392-0
CAS Number:7087-68-5
Formula:C₈H₁₉N
MDL Number:MFCD00008868