Magnesium chloride 0.01 mol concentrated aqueous solution volumetric solution
Catalog # CHMP166120500.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:95.21 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106