Order Entry
Poland
ContactUsLinkComponent
 
n-Hexadecane-[D₃₄] (98% D), Sigma-Aldrich®
Catalog # SIAL489603-100MG
Supplier:  Merck
CAS Number:  
n-Hexadecane-[D₃₄] (98% D), Sigma-Aldrich®
Catalog # SIAL489603-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
EINECS:208-878-9
 :GHS08
Storage Temperature:Ambient
CAS Number:15716-08-2
Formula:D₃C(CD₂)₁₄CD₃