Dinonyl phthalate (mixture of isomers) 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84-76-4
:
Formula:C₂₆H₄₂O₄
MDL Number:MFCD00036237
:418.62 g/mol
Density:0.97
Flash Pt:216 °C (421 °F)
Storage Temperature:Ambient
EINECS:201-560-0
Related Information
Frequently Bought Together